B21416.06
B21416.06EA
83.7
EUR
InStock
B21416.06
4,6-Dichloro-2-(methylthio)pyrimidine ≥98%
4,6-Dichloro-2-(methylthio)pyrimidine
Purity: 98%
CAS No.: 6299-25-8
Molecular Formula: C5H4Cl2N2S
Formula Weight: 195.07
Flash Point: 110°(230°F)
Odour: Irritating
Boiling Point: 135-136°/14mm
Beilstein Reference: 127664
UN No.: 1759
MDL No.: MFCD00006086