Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC) 0.10 %
B21551.14
B21551.14EA
209
EUR
InStock
B21551.14
2-Chlorostyrène ≥97% stabilisé
2-Chlorostyrène
Stabilized with 0.1% 4-tert-butylcatechol
Purity: 98%
CAS No.: 2039-87-4
Molecular Formula: C8H7Cl
Formula Weight: 138.60
Storage Temperature: 2°C to 8°C
Flash Point: 65°(149°F)
Refractive Index: 1.5650
Odour: Pungent
Melting Point: -63°
Boiling Point: 189°
Beilstein Reference: 2038863
MDL No.: MFCD00000567