B22001.06
B22001.06EA
96.6
EUR
InStock
B22001.06
2,4,6-Tris(trifluorométhyl)-1,3,5-triazine ≥98%
2,4,6-Tris(trifluorométhyl)-1,3,5-triazine
Purity: 98%
CAS No.: 368-66-1
Molecular Formula: C6F9N3
Formula Weight: 285.07
Flash Point: None
Refractive Index: 1.3220
Boiling Point: 95-96°
Beilstein Reference: 302339
MDL No.: MFCD00042436