Synonyms:
2,3,4,5,6-Pentafluorobenzyl cyanide, 2-(Perfluorophenyl)acetonitrile, Pentafluorobenzyl cyanide, (2,3,4,5,6-Pentafluorophenyl)acetonitrile
B22239.06
B22239.06EA
131
EUR
InStock
B22239.06
(Pentafluorophényl)acétonitrile ≥98%
(Pentafluorophényl)acétonitrile
Purity: 98%
CAS No.: 653-30-5
Molecular Formula: C8H2F5N
Formula Weight: 207.10
Flash Point: 110°(230°F)
Refractive Index: 1.4390
Boiling Point: 110-111°/17mm
Beilstein Reference: 1882341
UN No.: 3276
MDL No.: MFCD00013814