B22444.36
B22444.36EA
549
EUR
InStock
B22444.36
Acide 2-[(2-hydroxybenzoyl)oxy]benzoïque 98%
Acide 2-[(2-hydroxybenzoyl)oxy]benzoïque
Purity: 98%
CAS No.: 552-94-3
Molecular Formula: C14H10O5
Formula Weight: 258.23
Melting Point: 132-138°
MDL No.: MFCD00020252