Synonyms:
N,9-Diacetylguanine, 2-Acetamido-9-acetyl-6-oxopurine, N-(9-Acetyl-6-oxo-6,9-dihydro-1H-purin-2-yl)acetamide
B22709.06
B22709.06EA
32.4
EUR
InStock
B22709.06
N2,9-Diacetylguanine 99%
N2,9-Diacetylguanine
Purity: 99%
CAS No.: 3056-33-5
Molecular Formula: C9H9N5O3
Formula Weight: 235.20
Storage Temperature: -30°C to -10°C
Melting Point: 284-286°
MDL No.: MFCD00142116