B22939.22
B22939.22EA
55.7
EUR
InStock
B22939.22
Acide phénylphosphonique ≥98%
Acide phénylphosphonique
Purity: 98%
CAS No.: 1571-33-1
Molecular Formula: C6H7O3P
Formula Weight: 158.09
Melting Point: 160-162°
Beilstein Reference: 2245168
UN No.: 3261
MDL No.: MFCD00002136