B22962.AC
B22962.ACEA
26.5
EUR
InStock
B22962.AC
sec-Butylbenzène ≥99%
sec-Butylbenzène
Purity: 99%
CAS No.: 135-98-8
Molecular Formula: C10H14
Formula Weight: 134.22
Flash Point: 52°(125°F)
Refractive Index: 1.4900
Odour: Aromatic
Melting Point: -75°
Boiling Point: 173-174°
Beilstein Reference: 1903902
Merck Reference: 14,1550
UN No.: 2709
MDL No.: MFCD00009329