B23433.30
B23433.30EA
48.7
EUR
InStock
B23433.30
4-Éthylphénol ≥97%
4-Éthylphénol
Purity: 97%
CAS No.: 123-07-9
Molecular Formula: C8H10O
Formula Weight: 122.17
Flash Point: 100°(212°F)
Refractive Index: 1.5330
Melting Point: 40-45°
Boiling Point: 218-219°
Beilstein Reference: 1363317
UN No.: 2430
MDL No.: MFCD00002393