Synonyms:
4-Iodo-1,1,1-trifluorobutane, 1,1,1-Trifluoro-4-iodobutane
Stabilization:
Stabilised with copper
B23811.03
B23811.03EA
46.5
EUR
InStock
B23811.03
1-Iodo-4,4,4-trifluorobutane ≥98% stabilisé
1-Iodo-4,4,4-trifluorobutane
Stabilized with copper
Purity: 98+%
CAS No.: 461-17-6
Molecular Formula: C4H6F3I
Formula Weight: 237.99
Flash Point: 100°(212°F)
Refractive Index: 1.4335
Boiling Point: 126-127°
Beilstein Reference: 1736064
MDL No.: MFCD00041543