B23837.14
B23837.14EA
61.2
EUR
InStock
B23837.14
Cyclohexanecarboxaldehyde 97%
Cyclohexanecarboxaldehyde
Purity: 97%
CAS No.: 2043-61-0
Molecular Formula: C7H12O
Formula Weight: 112.17
Flash Point: 40°(105°F)
Refractive Index: 1.4500
Melting Point: 34-35°
Boiling Point: 161-163°
Beilstein Reference: 878306
UN No.: UN1989
MDL No.: MFCD00001457