B23906.22
B23906.22EA
24
EUR
InStock
B23906.22
Symclosène ≥90%
Symclosène
Purity: 90+%
CAS No.: 87-90-1
Molecular Formula: C3Cl3N3O3
Formula Weight: 232.41
Flash Point: 250°(482°F)
Odour: Pungent
pH: 3
Melting Point: ca 245° dec.
Beilstein Reference: 202022
Merck Reference: 14,9641
UN No.: 2468
MDL No.: MFCD00006553