B24385.30
B24385.30EA
73.8
EUR
InStock
B24385.30
1-(Chlorométhyl)naphtalène ≥94%
1-(Chlorométhyl)naphtalène
Purity: 94%
CAS No.: 86-52-2
Molecular Formula: C11H9Cl
Formula Weight: 176.65
Flash Point: 132°(270°F)
Refractive Index: 1.6370
Odour: Pungent
Melting Point: 31-33°
Boiling Point: 291-292°
Beilstein Reference: 636885
UN No.: 3261
MDL No.: MFCD00004042