Synonyms:
Valeric acid methyl ester, Methyl Pentanoate
B24828.22
B24828.22EA
34.3
EUR
InStock
B24828.22
Valérate de méthyle ≥99%
Valérate de méthyle
Purity: 99%
CAS No.: 624-24-8
Molecular Formula: CH3(CH2)3CO2CH3
Formula Weight: 116.16
Flash Point: 22°(71°F)
Refractive Index: 1.3970
Melting Point: -91°
Boiling Point: 126-128°
Beilstein Reference: 1741905
UN No.: 3272
MDL No.: MFCD00009478