Synonyms:
Monobromobimane, 3-(Bromomethyl)-2,5,6-trimethylpyrazolo[1,2-a]pyrazole-1,7-dione
M-Clarity™ quality level = MQ200
B4380-25MG
B4380-100MG
SIALB4380-25MGEA
156.5
EUR
InStock
SIALB4380-25MG
B4380-100MG
Bromobimane (mBBr) ≥97%, Sigma-Aldrich®
Bromobimane (mBBr)
Bromobimane is also known as monobromobimane. It is also a known probe for thiols and is a fluorescent reagent activated upon a photolysis reaction.