Sonorex Technik Ultrasonic Bath for Aqueous Fluids
Fournisseur: BANDELIN-ELECTRONIC
9233
BAND9233EA 0 EUR InStock
BAND9233
Sonorex Technik Ultrasonic Bath for Aqueous Fluids
Bains à ultrasons
SONOREX TECHNIK ultrasonic baths are intended for the sonication of aqueous liquids. They work on the basis of low-frequency ultrasound and can be used in versatile ways.
  • Designed as floor-standing units
  • Easy-to-clean stainless steel housing
  • Filling level mark for safe filling
  • Stainless steel oscillating tank with rounded tank corners and high-grade PZT
  • High-performance ultrasonic systems
  • Sprinkle tube and weir/overflow with ball valve to produce a liquid circuit
  • Height-adjustable legs for a secure and level position
  • Interference-free
Outlet with three way ball valve for easy draining/filling of the bath fluid. Their main application is the gentle and intensive cleaning of objects of diverse shapes, types and sizes. Tank: Stainless steel, 2 mm, welded, with weir on the right-hand side and sprinkle tube for surface skimming on the left-hand side of the tank.
Cable length: 3 m
Frequency: 40 or 25 kHz
Filling volume: 200 L
Operating volume: 170 L
Inlet and outlet: G 1 outside
Sprinkle tube inlet: G¹/₂ inside
Weir outlet: G 1 outside
Mains supply: 230 V~(±10%) 50/60 Hz for TM (U/UL); 400 V 3 N~ (±10%) 50/60 Hz for TM (UH/UHL)
Certifications: EN 61010-1, EMC: EN 61326-1, DIN EN 60529.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité