Digital heating/cooling block heater
Fournisseur: Thermo Fisher Scientific
88880030
BARN88880030EA 1640 EUR InStock
BARN88880030
Digital heating/cooling block heater
Chauffe-blocs Bains chauffants/réfrigérants
Compact programmable heating/cooling block heater with digital display can be used for all common applications in laboratory. Applications include enzyme reactions, transformation, DNA extractions, reactions that need to processed at or near room temperature and warming and cooling of reagents.
  • PID control ensures consistent precise temperature control
  • Stores 10 programs and 10 steps per program
  • Cord set with various plug types
  • Low-profile design
  • Blocks easily interchanged
Informations de commande: Blocks must be ordered separately.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité