Super Nuova+™ Hotplate Stirrer
Fournisseur: Thermo Fisher Scientific
SP88857195 SP88857198 SP88850195
BARNSP88857195EA 1120 EUR InStock
BARNSP88857195 BARNSP88857198 BARNSP88850195
Super Nuova+™ Hotplate Stirrer
Agitateurs Agitateurs magnétiques chauffants
Thermo Scientific™ SuperNuova+™ stirring hotplate performs with dependable safety and simple operation, units are available in two sizes including a multi-position model.
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for heating and stirring
  • Raised display design protects electronics from spills
  • Ability to set programs (ramp/dwell) as well as save settings
  • Optional PT1000 temperature probe and splash guard protection shield
Stir trac features for slow-speed stirring, consistent speed control, and stronger magnetic coupling. Hot Top warning system protects from accidental burns with prominent display when heating surface is above 50 °C (122 °F).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité