Serum separator tubes, SST™ II advance, BD Vacutainer®
Fournisseur: BD Medical
367957 367955 367953 367954 367956 367958 368879 366882 368969 368965 368966 368970
BDAM367957EA 38.3 EUR InStock
BDAM367957 BDAM367955 BDAM367953 BDAM367954 BDAM367956 BDAM367958 BDAM368879 BDAM366882 BDAM368969 BDAM368965 BDAM368966 BDAM368970
Serum separator tubes, SST™ II advance, BD Vacutainer®
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons de sang
PET, clear.
  • Silica (Clot Activator)/Gel
  • Stable barrier between serum and clotted blood, therefore better analyte stability
  • Better sample quality
  • Optimisation of the work flow: Short centrifugation time, sample processing and archiving in the primary tube
  • No possibility of misidentification due to the use of secondary tubes
During the centrifugation of the BD Vacutainer® SST™ II Advance tubes, an inert gel separates the serum and the blood clot preventing the contamination of the serum from the separated cellular components. For example, the serum for certain analytes such as potassium, phosphorus and glucose must be separated from the cells within a few hours - otherwise the results will be significantly distorted. Using BD SST™ II Advance tubes routine analytes in clinical chemistry such as potassium and glucose are still stable after a week of storage at 2-8ºC. Clinical evaluations of special chemistry demonstrate a high degree of analyte stability with the acrylic gel in the BD SST™ II Advance, with detection of >90% of the therapeutic drugs and other special analytes (proteins/peptides, steroids and vitamins) tested.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité