BD Nexiva™ Closed IV Catheter System
Fournisseur: BD Medical
383512
BDAM383512EA 501 EUR InStock
BDAM383512
BD Nexiva™ Closed IV Catheter System
Cathéters
The BD Nexiva™ closed IV catheter system is the integrated IV catheter demonstrating longer dwell times and reduced complication rates versus the traditional open system.
  • Proprietary BD Vialon™ biomaterial softens up to 70% in the vein, enabling longer dwell times and reducing the chance of mechanical phlebitis by up to 50%
  • BD Instaflash™ needle technology provides quick blood visualisation that may help improve insertion success and therefore reduce insertion attempts
  • Pre-attached extension tubing offers a closed system to minimise blood exposure during catheter insertion meeting the INS standards
  • 98% reduced blood exposure during insertion due to the BD Nexiva IV catheter pre-assembled systems
  • By preserving sites for longer, the BD Nexiva IV catheter helps patients get the medication they need as scheduled, potentially decreasing their length of stay
BD Nexiva™ IV catheter integrates key features that help provide better care and experience for your patients, efficiently and cost effectively.
Max power injector flow rate: 3 ml/sec
Max pressure limit: 300 psi
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité