BD® Arterial Cannula
Fournisseur: BD Medical
682245
BDAM682245EA 98.6 EUR InStock
BDAM682245
BD® Arterial Cannula
Canules
Designed specifically for arterial cannulation, the BD® arterial cannula enables arterial pressure monitoring and serial arterial blood gas determinations.
  • Developed to support smooth skin and vein penetration
  • Clear visibility of blood flashback confirms arterial entry
  • Flexible wings stabilise the catheter in place, helping to prevent unwanted movement
Device is designed to prevent back-flow, which potentially reduces the risk of air emboli and exposure to blood.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité