Serial extraction unit, round flasks, R 256 S, behrotest®
Fournisseur: BEHR
B00218436
BEHRB00218436EA 4920 EUR InStock
BEHRB00218436
Serial extraction unit, round flasks, R 256 S, behrotest®
Extracteurs Extracteurs Soxhlet
Behr equipment for Soxhlet extraction comply with the most various requirements of daily laboratory routines.
  • Practical holders for condenser and extraction adapter for secure placement between extractions
  • Extractor sizes from 30 ml to 1000 ml
  • Condenser with screw connections
Complete compact extraction system with basic rack, heating device, holder, hoses and glassware (reaction flasks, extractor, Dimroth condenser for extraction). Infinitely variable heating regulation After the extraction cycle, the compact system removes the solvent, by means of a discharge valve on the extractor directly into the reservoir.
With behr hydrolysis units, acid digestion is possible before extraction (determination of total fat content according to Weibull and Stoldt).
Information de livraison: Includes sample package, extraction sleeves and boiling stones.
Commander maintenant
COMMANDE
SPÉCIFICATIONS
ACCESSOIRES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité