Buchner funnel trowel, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
368200118
BELA368200118EA 204 EUR InStock
BELA368200118
Buchner funnel trowel, SP Bel-Art
Entonnoirs Entonnoirs pour la filtration
Specifically designed for use with 18, 24 and 36” Buchner Funnels, apply slight downward pressure and ‘sweep’ the trowel along the surface of the precipitate to quickly and efficiently smooth fissures appearing during the filtration process.
  • Large surface area smooths fissures faster than the traditional narrow spatula method
  • Beveled edges on both sides permit bi-directional smoothing
  • Curved ends match Buchner Funnel sidewalls allowing easy access to the entire cake surface
  • FDA-grade high-density polyethylene (HDPE) complies with industry standards and provides maximum chemical-resistance
  • Welded construction with no hollow cavities assures hygienic use and easy cleaning
Compatible with industrial dishwasher temperatures and cleaners.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité