Incubator thermometer, DURAC® Plus™, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
B606000200
BELAB606000200EA 177 EUR InStock
BELAB606000200
Incubator thermometer, DURAC® Plus™, SP Bel-Art
Thermomètres Thermomètres analogiques
This mercury-free thermometer is designed to verify sample temperatures in incubators, and cover ranges from 10 to 50 °C.
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle (included), which mimics the sample
  • Features permanently fused markings and lead-free glass
  • Insert thermometer into the bottle and place near the samples for best results
Each instrument is individually calibrated and certified against equipment whose calibration is traceable to the National Institute of Standards and Technology.
Certifications: Traceable to NIST, individually serialised.
Information de livraison: Includes multi-language (English, French, German, Italian, Portuguese, and Spanish) statement of accuracy and traceability.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité