SP Bel-Art H-B DURAC® Calibrated Electronic Thermometer with Stainless Steel Probe
Fournisseur: Bel-Art Products, a Part of SP
B60900-3400
BELAB60900-3400EA 0 EUR InStock
BELAB60900-3400
SP Bel-Art H-B DURAC® Calibrated Electronic Thermometer with Stainless Steel Probe
Thermomètres Thermomètres numériques
Monitor the temperature of pharmaceuticals, vaccines, chemicals, blood and blood products, food products and more with these high accuracy digital thermometers with chemical resistant probes.
  • Features available include 10-event recording, minimum/maximum temperature readings, timers and alarms
  • All models include accredited calibration documentation that meets the requirements of ISO 17025
  • Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish)
  • Individually serialized
Tested and calibrated in H-B's exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité