Wash bottles, wide neck, right-to-know, safety vented, with GHS labelling, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
F124160012
BELAF124160012EA 143 EUR InStock
BELAF124160012
Wash bottles, wide neck, right-to-know, safety vented, with GHS labelling, SP Bel-Art
Flacons Pissettes
Translucent LDPE bottles with color-coded caps for easy identificatio.
  • Wide mouth 53 mm for easy refilling
  • Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word
  • Methyl Ethyl Ketone and Toluene do not include vented valve
Specially designed valve prevents pressure build-up that eliminates potentially dangerous chemical drips; allows dispensing of wash bottle in upright or inverted position. Pre-printed with chemical name and formula, GHS pictogram and signal word, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.
Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word.
Certifications: Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA's HazCom 2012 requirements–as pertains to 'Workplace Labels'
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité