Mini cryocooler, –15 °C, Bel-Art ProCulture, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
F188460020
BELAF188460020EA 306 EUR InStock
BELAF188460020
Mini cryocooler, –15 °C, Bel-Art ProCulture, SP Bel-Art
Boîtes Boîtes de congélation
Polycarbonate cooler body gel-filled with clear polycarbonate lid.
  • Adapters change well size to stow 0,5 ml microcentrifuge tubes
  • Small size is ideal for benchtop use
  • All polycarbonate cooler body cleans easily
  • Use to protect samples in freezers with defrost cycle and fluctuating temperatures
A non-toxic freezing gel is isolated within the walls of the cooler, guaranteeing that any labels or marks on the vials or tube are not damaged. The indexed and transparent polycarbonate lid shows tube location and matches locations printed on the cooler. There are side handles for easy carrying. Coolers can be stacked to save space and maximize freezer volume and have rubber feet to prevent slipping. Stow twelve 1,5 ml microcentrifuge tubes below 15 °C for roughly 1 hour.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité