Cryo Storage Box −20 °C Cryo-Safe™ Maxi Cooler, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
F188462020
BELAF188462020EA 490 EUR InStock
BELAF188462020
Cryo Storage Box −20 °C Cryo-Safe™ Maxi Cooler, SP Bel-Art
Boîtes Boîtes de congélation
Polycarbonate cooler body and lid, gel-filled.
  • Adapters change well size to hold 0,5 ml microcentrifuge tubes
  • Small footprint is ideal for benchtop use
  • Epoxy coated wire handle fastens lid and allows easy transport
  • All polycarbonate cooler body and lid clean easily
A non-toxic freezing gel is isolated within the walls of the cooler and lid, ensuring lasting low temperatures and protecting any labels or marks on the vials. The lid is indexed and marked to show tube location that matches locations printed on the cooler. Coolers can be stacked to save space and maximize freezer volume and have rubber feet to prevent slipping. Ideal for protecting samples in freezers with defrost cycle and fluctuating temperatures. Keep thirty-two 1,5 ml microcentrifuge tubes below −20 °C for 1,5 hours on the benchtop.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité