Sterileware double-ended pestle, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
F199110001
BELAF199110001EA 90.7 EUR InStock
BELAF199110001
Sterileware double-ended pestle, SP Bel-Art
Mortiers et pilons Pilons
Double-ended pestle has flat ends to effectively press soft tissues through screens, thereby disrupting the organ structure and producing cell isolates.
  • Double-ended design allows users to choose preferred pestle diameter
  • Chemical-resistant and autoclave-safe FDA-grade polypropylene construction
  • Individually packaged and gamma sterilised and for convenience
  • End Ø: 0,95 cm and 1,27 cm
Sterilised products are not returnable.
Information de livraison: 12-well tissue disaggregator includes one stainless steel autoclave frame and 12 individually packaged sterile pestles.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité