Micro spoon and spatula sampling set, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
F367050000
BELAF367050000EA 265 EUR InStock
BELAF367050000
Micro spoon and spatula sampling set, SP Bel-Art
Spatules
This kit includes six different, small size, double-ended tools and a pair of tweezers. Each double-ended tool has two different end shapes providing 12 implements to better match user needs. Kits include:
  • Two small spoons for transferring and collecting both dry and liquid material
  • Two V-shaped spatula ends of different sizes that can be used for lifting or spill-free carrying of material
  • Eight flat spatulas ends (six square ends and two rounded) of various widths and lengths ideal for mixing, scraping and spreading
  • Blunt tip tweezers have serrated inside surfaces for secure sample handling and grooved handles for better gripping
All tools are polished stainless steel for long life and easy cleaning and can be steam autoclaved at 121 °C. A carrying case protects and organises all tools when not in use.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité