Sampling spatulas, earth-friendly, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
F369310000
BELAF369310000EA 154 EUR InStock
BELAF369310000
Sampling spatulas, earth-friendly, SP Bel-Art
Spatules
PLA resin, translucent. These non sterile sampling sticks are made of earth-friendly polylactic acid.
  • Green alternative to petroleum-based sampling tools; helps meet sustainability goals
  • Comparable in strength and durability to PS, PP and other petroleum-based plastic resin sampling tools
  • Disposal options include industrial composting for biodegradability, incineration, recycling and landfill
  • Non sterile FDA Grade: safe for food, drug and cosmetic contact
The production of 100% plant-based PLA contributes less greenhouse gasses and uses less non renewable energy from source to resin than its petroleum-based counterparts, and the plants used are an annually renewable resource. The sticks are ideal for mixing samples, handling capsules, scraping containers or positioning solid samples for examination.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité