Battery powered engraving tool, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
F441500005
BELAF441500005EA 64.9 EUR InStock
BELAF441500005
Battery powered engraving tool, SP Bel-Art
Stylos Stylets et traceurs
Battery powered engraving tool to permanently mark most materials including glass, plastic, wood, metal, bone, and stone.
  • Cordless battery power makes it portable for use virtually anywhere indoors or out in the field
  • Marks will not fade if exposed to chemicals or sunlight
  • Operates on two 'AA' batteries
Simply hold the engraving tool like a pen, press the power button, apply light pressure and write on test tubes, bottles, racks, keys, equipment, tools, electronics and more.
Informations de commande: Convenient polyethylene stand (sold separately)More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité