Tape dispensers for labelling, single roll, Write-on™, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
H134610000
BELAH134610000EA 240 EUR InStock
BELAH134610000
Tape dispensers for labelling, single roll, Write-on™, SP Bel-Art
Distributeurs d'étiquettes et de rubans adhésifs
Designed for one hand label making, the dispenser comes with a weighted bottom with non-skid pads to avoid sliding or lifting. The epoxy-coated steel platform has a jagged edge and finger holes for releasing the label tape. The writing platform accommodates left and right-handed users. Dispenser accepts any tape with a 7,6 cm core and maximum OD of 133 mm. Ideal for use with Write-On™ Label Tape. Dimensions: 213× 87×95 mm, platform: 102×152 mm, shipping weight: 1,8 kg.
  • Quickly produce neat stick-on labels with just one hand
  • Fresh piece of label tape is always positioned on the flat, smooth writing platform
  • Holds tape up to 25 mm wide
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité