Support stand with rod, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
H183030000
BELAH183030000EA 272 EUR InStock
BELAH183030000
Support stand with rod, SP Bel-Art
Supports de statif
This heavy-duty support stand is made of PP plastic and metal, yet has no exposed metal parts. Excellent for use in severe corrosive conditions.
  • Base has a smooth surface for easy cleaning and weighs 2,5 kg for stability
  • Rod (12,5×760 mm) can be screwed into the center of the base, or at either end
  • Rod has a deep thread and sturdy tightening knob
  • Autoclavable
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité