Fume hood, benchtop, small, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
H500000002
BELAH500000002EA 1070 EUR InStock
BELAH500000002
Fume hood, benchtop, small, SP Bel-Art
Hottes et coffrets Sorbonnes pharmaceutiques
Benchtop fume hood designed for small facilities or installations in which a number of fume hoods are needed, this unit is mainly used in schools, junior colleges and universities.
  • Easy to clean
  • Bottom gap allows for continuous air flow
  • One-piece shell moulded of chemical resistant polyethylene with rounded inside corners
  • Clear acrylic sash slides on a track featuring stop pins for open, closed or intermediate positions
Commander maintenant
COMMANDE
SPÉCIFICATIONS
SERVICES
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité