SenTix® pH Glass Electrodes with Temperature Sensor
Fournisseur: BELLINGHAM & STANLEY
103810
BELL103810EA 298 EUR InStock
BELL103810
SenTix® pH Glass Electrodes with Temperature Sensor
Électrodes Électrodes pH
The SenTix® pH electrodes with their high-quality glass shaft and refillable 3 mol/L KCl liquid electrolyte and platinum diaphragm are suitable for demanding measurements in the laboratory. The platinum wire diaphragm offers optimum electrolyte flow without clogging and the liquid electrolyte offers fast response times. The electrodes can also be used in difficult samples. The cone shape of the membrane is impact-resistant and easy to clean.
  • Glass shaft and refillable KCl electrolyte
  • Platinum wire diaphragm for clog resistance
  • Integrated temperature sensor
  • Impact-resistant cone membrane for fast response
  • Multiple configurations with plug options
Chemical applications: Diluted acids and Emulsions/suspensions
Industrial applications: Cooling water, dyeing solutions, electroplating baths and Paper extract
Water and environmental applications: Saline solutions, swimming pool water, groundwater, lake water, surface water, sea water, drinking water and soil extract
Cosmetics and cleaning applications: Hair color, mouthwash and household cleaners
Food applications: Coffee extract and vinegar
Pharma, Medical, Bio applications: Enzyme solutions, gastric juice, infusion solutions, protein-containing liquids, serum, tris buffer solutions, urine
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité