Digital refractometer, RFM742-M
Fournisseur: BELLINGHAM & STANLEY
19-20
BELL19-20EA 9244 EUR InStock
BELL19-20
Digital refractometer, RFM742-M
Réfractomètres Réfractomètres de paillasse
The RFM742-M refractometers are robust fully automatic instruments suited to the food, sugar and beverage industries where temperature control is not required, such as measuring sugar (°Brix) concentrations in the food processing industry.
  • Automatic sense 'hands free' measurements
  • Simple audit trail: Date, time and batch number
  • Alpha-numeric keypad for easy data entry
  • Flat sapphire prism surface for easy-cleaning
  • Selectable read type: Continual, single, intelligent auto-sense
May be used in the traditional food, sugar and beverage applications, and in 'non-sucrose' based industrial applications.
Certifications: 'Fuel System Icing Inhibitor' scale that conforms to ASTM D 5006 can be loaded as well as refractive index (RI).
Information de livraison: Supplied with power cable and instruction manual. EU, UK and US plugs are supplied with the unit, CH and Danish plugs must be ordered separately.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité