Cleanroom wipes, CHOICE® 900
Fournisseur: BERKSHIRE
CH909.10
BERKCH909.10EA 526 EUR InStock
BERKCH909.10
Cleanroom wipes, CHOICE® 900
Lingettes Lingettes pour salles propres Lingettes pour salles propres, sèches
Cleanroom wipes made of 100% continuous filament knit polyester, ultrasonically sealed edge for reduced fiber contamination.
  • Critically low particles, fibers, ions and extractables
  • High abrasion resistant
  • Excellent absorbency
  • Chemically compatible with IPA, Acetone and other solvents
Designed for use in ISO Class 3 and higher environments.
Suitable for breakdown cleans in fab and suite construction, oxidation, metallisation or CVD deposition areas, chamber cleaning and CMP processing, stencil and other print roll cleaning applications, cleaning of medical device products, applying and removing cleaning and disinfecting solutions.
Certifications: Complies with RoHS and Reach requirements.
Conditionnement: 150 sheets per pack, 10 packs per case (double packed).
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité