EcoClean® 60 Cleanroom Cotton Wipes, Non woven
Fournisseur: BERKSHIRE
ECW60090920
BERKECW60090920EA 1350 EUR InStock
BERKECW60090920
EcoClean® 60 Cleanroom Cotton Wipes, Non woven
Lingettes Lingettes pour salles propres Lingettes pour salles propres, stériles
EcoClean® is an environmentally friendly wipe that has been designed for ISO Class 5 (Class 100) and above cleanroom applications. It is made from 100% natural fiber that contains no binders or additives, and it is manufactured by a proprietary process for low fiber release and high strength.
  • 100% nonwoven cotton
  • Manufactured by a proprietary process for low fiber release and high strength
  • No binders or other additives
  • Low ionic contamination
  • High strength
  • Heat resistant
  • Highly absorbent and durable
  • Extremely soft
  • Resists build up of static charge
Applications includes:
• Cleaning high temperature production equipment and diffusion furnaces
• Absorbent, low linting work surface material
• Removing aqueous and solvent spills
• Critical industrial applications where absorbency and softness are critical
• Cleaning of support/less critical areas and manufacturing processes in regulated environments
• Wiping of optics and other sensitive surfaces
• Steam autoclavable for aseptic applications
Certifications: Compatible with ISO Class 5 (Class 100) cleanroom applications and above, FDA 21 CFR 177.2800 (GRAS).
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité