Cleanroom wipes, MicroPolx® 4000-HP
Fournisseur: BERKSHIRE
MP4000HP090910
BERKMP4000HP090910EA 903 EUR InStock
BERKMP4000HP090910
Cleanroom wipes, MicroPolx® 4000-HP
Lingettes Lingettes pour salles propres Lingettes pour salles propres, sèches
These wipes made from 80% Polyester and 20% nylon micro denier knitted fabric are designed for the highest level of contamination control in critical processing applications.
  • Microfibre yarn increases surface area and provides superior wiping efficiency and sorptive performance
  • Cleans efficiently with less solvent usage than traditional wipes
  • Removes oily soils with little or no solvent required
  • Soft hand and non-abrasive sealed-edge are excellent for sensitive or scratch prone surfaces
  • Ultrasonically sealed edges minimise fibre and particle generation
  • Critically low particles, fibres, ions and extractables
  • High abrasion resistance
  • Chemically compatible with IPA and other solvents
Ideal for cleaning precise optical machinery and tools, semiconductors and for cleaning a delicate process in critical cleanrooms.
Laundered and packaged in an ISO Class 4 cleanroom. Suitable for use in Class 4 cleanrooms.
Attention: Data accurate at time of publishing, latest test results and test methods can be found on supplier's website.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité