ValuClean® Plus 50% Rayon/50% Polyester Spunlace Cleanroom Wiper
Fournisseur: BERKSHIRE
VCLP.0909.20
BERKVCLP.0909.20EA 700 EUR InStock
BERKVCLP.0909.20
ValuClean® Plus 50% Rayon/50% Polyester Spunlace Cleanroom Wiper
Lingettes Lingettes pour salles propres Lingettes pour salles propres, sèches
ValuClean® Plus is a hydroentangled rayon/polyester nonwoven wiper recommended for ISO class 5 and above environments and offers durability and cleanliness.
  • Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of
  • rayon
  • Smooth with excellent strength and durability
  • High absorbency compared to typical cellulose/polyester nonwoven wipers can reduce actual wiper usage in
  • spill or solution removal applications
  • Low extractable levels and particle counts
  • Chemically compatible with common cleaning and disinfecting solutions
  • Economical
Pre wetted option: The same wiper material can be provided in pre-wetted formats for reduced VOC emissions, increased convenience, increased productivity, improved solvent control and cleaning protocol repeatability and reduced costs. Sterile Validated Option: For aseptic processing areas, the same wiper material can be provided in a gamma irradiated validated sterile to a 10⁻⁶ sterility assurance level. Dry and pre-wetted versions available.
Low basis weight reduces landfill waste impact when compared with market leading cellulose/polyester wipers. General wiping in component prep, compounding and wash areas, applying and removing cleaning and disinfecting solutions and spill pick up, general wipe downs, and cleaning of lab equipment.
Due to 50% rayon content, ValuClean® Plus has absorbency reducing actual wiper usage in spill removal applications. Its soft hand and low fiber shedding performance make it a good choice in critical cleaning applications
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité