Cleanroom wipes, sterile, sealed edge, ValuSeal® IonX™
Fournisseur: BERKSHIRE
VSI.0909.14
BERKVSI.0909.14EA 805 EUR InStock
BERKVSI.0909.14
Cleanroom wipes, sterile, sealed edge, ValuSeal® IonX™
Lingettes Lingettes pour salles propres Lingettes pour salles propres, sèches
Designed for the highest level of contamination control in critical processing applications.
  • Critically low particles, fibres, ions, and extractables
  • High abrasion resistance
  • Lightweight, high absorbency material reduces actual cost in use compared to heavier weight products
  • Laser sealed edge for reduced fibre contamination
ValuSeal® IonX® is a laser sealed edge cleanroom laundered wipe recommended for ISO Class 3 and above environments. Composed of 100% polyester continuous filament yarns, its smooth knit construction and laser sealed edges make it ideal for applications requiring critically low levels of particle and fibre release, along with durability and chemical compatibility.
Certifications: ISO Class 3 – 5. Cleanroom Class 1 – 100. EU Grade B/C/D.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité