Water treatment system, Pure Aqua Service
Fournisseur: Binder
8012-0759
BIND8012-0759EA 1281 EUR InStock
BIND8012-0759
Water treatment system, Pure Aqua Service
Purificateurs d'eau Déioniseurs
This system is used to fully desalinate faucet water for BINDER climate chamber humidification systems. The ion exchanger system consists of a pressure container, a single-use filter cartridge and a pressure container lid.
  • Can be used irrespective of drinking water quality
  • Robust system with safety pressure container
  • Reliable replacement of single-use cartridge when resin is used up
  • Operating condition of water treatment system is permanently displayed
The continuous water quality monitoring system is integrated in the lid of the Pure Aqua Service andMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité