Disarticulated human skeleton
Fournisseur: 3B Scientific
1000026
BINH1000026EA 0 EUR InStock
BINH1000026
Disarticulated human skeleton
Modèles Modèles de biologie humaine
This disarticulated skeleton is hand-painted and numbered to show muscle origins in red and muscle insertions in blue on the left side. Bones and bony structures such as fissures, foramina and processes are hand-numbered on the right side. The skull is detachable into three parts for detailed anatomical study. The muscle disarticulated full skeleton features:
  • One hand and foot on wire
  • One set of arms and leg loosely
  • Multilingual product manual to identify over 600 structures
Information de livraison: Supplied in a sturdy partitioned cardboard storageMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité