Painted human skull model, 3 parts
Fournisseur: Binhold, Paul Lehrmittel
1000055
BINH1000055EA 0 EUR InStock
BINH1000055
Painted human skull model, 3 parts
Modèles Modèles de biologie humaine
The muscle origins (red) and insertions (blue) are shown in color on the left side of the skull. Cranial bones and structures are numbered on the right side. This skull model identifies over 140 anatomical details:
  • High-quality original casts
  • Handmade of hard, unbreakable plastic
  • Highly accurate representation of the fissures, foramina, processes, sutures
  • Can be disassembled into skull cap, base of skull and mandible
  • As an option, you can insert a five-part brain (C 18) into the skull
This replica of the human skull is designed for teaching and learning the anatomy of the skull.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité