Beauchene adult human skull model
Fournisseur: Binhold, Paul Lehrmittel
1000069
BINH1000069EA 0 EUR InStock
BINH1000069
Beauchene adult human skull model
Modèles Modèles de biologie humaine
Beauchene adult human skull is a natural cast and makes the complexity of the anatomical structure easy to understand, since it is disassembled into its 22 individual bones.
  • Bones/bone pairs are coloured to make dentification easier
  • Bones can be reassembled by means of inconspicuous, stable connectors attached at the slightly simplified skull sutures
  • Human skull consists of individual bones that gradually grow together as the development proceeds
This skull model is a great tool for teaching and learning the complex anatomy of the human skull.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité