Suture practice leg
Fournisseur: Binhold, Paul Lehrmittel
1005683
BINH1005683EA 462 EUR InStock
BINH1005683
Suture practice leg
Modèles Modèles de biologie humaine
Made with a soft vinyl skin over a core of stitchable foam to provide a lifelike suturing experience for students or a realistic suturing demonstration by an instructor. Realistic skin texture with wrinkles and pores. Soft and pliable for easy sewing, the skin is tough enough that sutures will not pull out when tightened. Each unit provides three 'wounds'. These 'wounds' can be sutured repeatedly until the skin around them is finally worn out; then, new 'wounds' can be cut. Although disposable, the trainers provides hundreds of suturing experiences before wearing out. Conservatively, over one hundred cuts can be made on the trainer and each of these cuts can be sutured several times.
  • Complete with a starter suturing kit and instruction booklet
  • Dimensions: 62×18×13 cm
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité