Skeleton model with painted muscle origins and inserts - Max
Fournisseur: Binhold, Paul Lehrmittel
1013858
BINH1013858EA 0 EUR InStock
BINH1013858
Skeleton model with painted muscle origins and inserts - Max
Modèles Modèles de biologie humaine
Max has all the advantages of a 3B Scientific® skeleton and also shows the structural interaction between the bones and muscles. On the left side of the skeleton over 600 important medical and anatomical structures are shown, such as the muscle origins (red) and insertions (blue) as well as hand-numbered bones, fissures and foramina on the right side. The advantages of the 3B Scientific® skeleton are:
  • Exceptional value for money
  • 3 year guarantee
  • Top quality natural casting 'Made in Germany'
  • Final assembly carried out by hand
  • Made from a durable, unbreakable synthetic material
  • On a stable metal stand with 5 casters (painted white)
  • Close to the realistic weight of around 200 bones
  • Natural size
  • Anatomically correct
  • 3 part assembled skull
  • Individually inserted teeth
  • Limbs can be removed quickly and easily
  • With transparent dust cover
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité