Electron diffraction tube D
Fournisseur: Binhold, Paul Lehrmittel
1013885
BINH1013885EA 1020 EUR InStock
BINH1013885
Electron diffraction tube D
Matériel éducatif Matériel éducatif de physique Activités d'électricité/de magnétisme
Highly evacuated electron tube for determining the wavelength of electrons as a function of the anode voltage from the radii of the diffraction rings and the distance between the lattice planes of graphite (Debye-Scherrer-diffraction) and rendered visible on a fluorescent screen. Also intended for determining the wavelengths of monochromatic radiation at different anode voltages from the radii of the diffraction rings and the lattice plane spacing of graphite, as well as confirming de Broglie’s hypothesis.
  • Lattice constant of graphite: d10 = 0,213 nm, d11 = 0,123 nm
Filament voltage: 6,3 V AC; Max. anode voltage: 5000 V; Focussing voltage: 0 to 50 V
Informations de commande: Power supplies, connection leads and tube holder need to be ordered separately, please ask for details.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité