Human MIA ELISA Kit
Fournisseur: Biorbyt
ORB106600-96
BIRBORB106600-96EA 0 EUR InStock
BIRBORB106600-96
Human MIA ELISA Kit
Tests Tests ELISA
Human MIA ELISA Kit for the quantitative detection of human Melanoma Inhibiting Activity (MIA).
Melanoma Inhibiting Activity (MIA), also known as cartilage-derived retinoic acid-sensitive protein (CD-RAP), is an approximately 11-15 kDa protein that is expressed as a noncovalent homodimer. MIA is structurally related to OTOR/Otoraplin and MIA-2 in a small family of secreted proteins with one SH3 domain (1-3). And the MIA gene was mapped to 19q13.32-q13.33 by fluorescence in situ hybridisation. Beside, It is a marker for malignant melanoma. MIA functions as a chemoattractant for mesenchymal stem cells and enhances their BMP-2 and TGF-3 induced differentiation into chondrocytes. MIA-deficient mice exhibit delayed chondrocyte differentiation but enhanced chondrocyte proliferation and cartilage repair.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité